missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Syk (Tyr323) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538335
Additional Details : Weight : 0.00900kg
Description
Syk (spleen tyrosine kinase) is a 635 amino acid protein that contains one protein kinase domain and two SH2 domains. One of several members of the protein kinase superfamily, Syk functions as a positive effector of B cell antigen receptor (CD79)-stimulated responses, coupling CD79 with the movement of one calcium ion through one of two phospho-regulated pathways. Via its ability to influence CD79 activity and to control the movement of calicum through the cell, Syk plays an important role in a variety of cellular responses, including differentiation, phagocytosis, proliferation and B cell development. Syk expression is upregulated in T cell lymphoma, suggesting a possible role for Syk in tumorigenesis. Two isoforms of Syk, designated short and long, exist due to alternative splicing events.Specifications
Phospho-Syk (Tyr323) | |
Polyclonal | |
Unconjugated | |
SYK | |
Tyrosine-protein kinase SYK, Spleen tyrosine kinase, p72-Syk, SYK | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20963, 25155, 6850 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P43405, P48025, Q64725 | |
SYK | |
A synthetic phosphopeptide derived from human SYK around the phosphorylation site of Tyr323 (N-P-YP-E-P) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |