missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Vinculin (Tyr822) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 441080G
Additional Details : Weight : 0.50000kg
Description
Phospho-Vinculin (Tyr822) Polyclonal antibody specifically detects Phospho-Vinculin (Tyr822) in Chicken, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-Vinculin (Tyr822) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P12003, P18206, P85972, Q64727 | |
VCL | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human vinculin that contains tyrosine 822. The sequence is conserved in mouse, rat, and chicken. | |
100 μL | |
Primary | |
Chicken, Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
VCL | |
9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; metavinculin; meta-vinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; unnamed protein product; Vcl; vcl {ECO:0000250; VIN; vinc; vinc 1; VINC1; Vinculin; vinculin {ECO:0000250 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22330, 305679, 396422, 7414 | |
-20°C | |
Liquid |