Läs mer
PGK1 Monoclonal Antibody (22C5D8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 459250
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This product reacts with Saccharomyces cerevisiae PGK1 - predicted molecular weight: 45 kDa. Purity >95% by SDS-PAGE.
PGK1 Monoclonal antibody specifically detects PGK1 in Yeast samples. It is validated for Western Blot.
Specifikationer
| PGK1 | |
| Monoclonal | |
| 1 mg/mL | |
| HEPES buffered saline with 0.02% sodium azide | |
| cell migration-inducing gene 10 protein; epididymis secretory sperm binding protein Li 68p; HEL-S-68p; MIG10; OK/SW-cl.110; Pgk; Pgk1; Pgk-1; PGKA; Phosphoglycerate kinase; phosphoglycerate kinase 1; Primer recognition protein 2; PRP 2; YCR012W; YCR12W | |
| Mouse | |
| purified | |
| RUO | |
| 850370 | |
| 4° C | |
| Liquid |
| Western Blot | |
| 22C5D8 | |
| Unconjugated | |
| PGK1 | |
| PGK1 | |
| Full-length native S. cerevisiae pGK protein (purified) | |
| 250 μg | |
| Primary | |
| Yeast | |
| Antibody | |
| IgG1 κ |