Läs mer
PR domain containing 1, with ZNF domain, Mouse, Clone: 2F8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PRDM1.
Brand: Abnova H00000639-M04.100ug
Beskrivning
This gene encodes a protein that acts as a repressor of beta-interferon gene expression. The protein binds specifically to the PRDI (positive regulatory domain I element) of the beta-IFN gene promoter. Transcription of this gene increases upon virus induction. Two alternatively spliced transcript variants that encode different isoforms have been reported. [provided by RefSeq
Sequence: MKMDMEDADMTLWTEAEFEEKCTYIVNDHPWDSGADGGTSVQAEASLPRNLLFKYATNSEEVIGVMSKEYIPKGTRFGPLIGEIYTNDTVPKNANRKYFWRIYSRGELHSpecifikationer
| PR domain containing 1, with ZNF domain | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001198 | |
| PRDM1 | |
| PRDM1 (NP_001189, 1 a.a. ∼ 109 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2F8 | |
| Mouse monoclonal antibody raised against a partial recombinant PRDM1. | |
| PRDM1 | |
| BLIMP1/MGC118922/MGC118923/MGC118924/MGC118925/PRDI-BF1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 639 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.