missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG Fc Secondary Antibody, Biotin, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A16122
Additional Details : Weight : 0.01000kg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Rabbit IgG Fc Polyclonal antibody specifically detects Rabbit IgG Fc in Rabbit samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
Rabbit IgG Fc | |
Polyclonal | |
Biotin | |
Goat | |
Affinity chromatography | |
RUO | |
Rabbit | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobin Fc region | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |