missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ SEL1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524179
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
SEL1L Polyclonal Antibody for Western Blot, IHC (P), Flow
The protein encoded by this gene is part of a protein complex required for the retrotranslocation or dislocation of misfolded proteins from the endoplasmic reticulum lumen to the cytosol, where they are degraded by the proteasome in a ubiquitin-dependent manner. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifikationer
| SEL1L | |
| Polyclonal | |
| Unconjugated | |
| SEL1L | |
| AW493766; fj60e07; IBD2; mKIAA4137; PRO1063; Protein sel-1 homolog 1; Sel1 (suppressor of lin-12) 1 homolog; sel-1 suppressor of lin-12-like; sel-1 suppressor of lin-12-like (C. elegans); sel-1 suppressor of lin-12-like 1; Sel1h; SEL1L; sel-1L; SEL1L adaptor subunit of ERAD E3 ubiquitin ligase; SEL1L ERAD E3 ligase adaptor subunit; SEL1L, ERAD E3 ligase adaptor subunit; SEL1L1; SEL1-LIKE; si:dkey-177p2.3; Suppressor of lin 12 (sel-1), C. elegans, homolog of; suppressor of lin-12-like protein 1; TSA305; UNQ128/PRO1063; wu:fj60e07; zgc:56075 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 6400 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9UBV2 | |
| SEL1L | |
| KLH conjugated synthetic peptide between 156-182 amino acids from the central region of human SEL1L. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering