Learn More
SIRP alpha Monoclonal Antibody (OX-41), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA170021
1513.40 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
MA1-70021 detects SIRP in rat samples. MA1-70021 has been successfully used in flow cytometry, immunocytochemistry, immunofluorescence, and radioiummunoassay procedures. The MA1-70021 immunogen is rat peritoneal macrophages.
SIRP alpha Monoclonal antibody specifically detects SIRP alpha in Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Radioimmune Assays (RIA).Specifications
SIRP alpha | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P97710 | |
SIRPA | |
Rat peritoneal macrophages. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Radioimmune Assays (RIA) | |
OX-41 | |
Unconjugated | |
SIRPA | |
AI835480; Bit; brain Ig-like molecule with tyrosine-based activation motifs; Brain immunoglobulin like protein with tyrosine - based activation motifs; brain immunological-like with tyrosine-based motifs; brain-immunoglobulin-like molecule with tyrosine-based activation motifs; CD172; CD172 alpha; CD172 antigen-like family member A; CD172A; Inhibitory receptor SHPS-1; macrophage fusion receptor; Macrophage membrane protein MFP150; MFR; mSIRP-alpha1; Myd1; MYD-1; myD-1 antigen; p84; Protein tyrosine phosphatase non-receptor type substrate 1 (SHP substrate 1); protein tyrosine phosphatase, non-receptor type substrate 1; Protein tyrosine phosphatase, non-receptor type substrate 1 (SHP substrate 1); Ptpns1; SHP substrate 1; SHP-1; SHPS1; SHPS-1; signal regulatory protein alpha; signal-regulatory protein alpha; signal-regulatory protein alpha-1; Signal-regulatory protein alpha-2; Signal-regulatory protein alpha-3; SIRP; Sirpa; SIRPalpha; Sirp-alpha-1; SIRPalpha2; Sirp-alpha-2; Sirp-alpha-3; | |
Mouse | |
Protein G | |
RUO | |
25528 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |