missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMYD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519595
Additional Details : Weight : 0.01000kg
Description
Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with mouse and chicken based on sequence homology.
Specifications
SMYD1 | |
Polyclonal | |
Unconjugated | |
Q8NB12 | |
SMYD1 | |
Synthetic peptide conjugated to KLH derived from within residues 450 to the C-terminus of Human SMYD1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.7 mg/mL | |
SMYD1 | |
BOP, KMT3D, ZMYND18, ZMYND22, CD8 beta opposite; SET and MYND domain-containing protein 1; zinc finger, MYND domain containing 18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |