missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAIL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523482
Additional Details : Weight : 0.01000kg
Description
SNAIL Polyclonal antibody specifically detects SNAIL in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SNAIL | |
Polyclonal | |
Unconjugated | |
Snai1 | |
AI194338; dJ710H13.1; Protein sna; protein snail homolog 1; SLUGH2; SNA; Sna protein; Sna1; SNAH; snai; SNAI1; Snail; snail 1; snail 1 homolog; snail 1 zinc finger protein; snail 1, zinc finger protein; snail family transcriptional repressor 1; snail family zinc finger 1; snail homolog 1; Snail1; Zinc finger protein SNAI1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20613, 6615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
O95863, Q02085 | |
Snai1 | |
A portion of amino acids 1-60 of human SNAI1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |