missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAP25 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA19102
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine and rat based on sequence homology.
SNAP25 Polyclonal antibody specifically detects SNAP25 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SNAP25 | |
Polyclonal | |
Unconjugated | |
Snap25 | |
ALMS1; bA416N4.2; Bdr; blind drunk; CG17676; CG17884; CG40452; CMS18; dJ1068F16.2; Dmel CG40452; Dmel_CG40452; etID17571.20; etID69167.20; fj42b02; fj45g04; fj48c02; FLJ23079; GENA70; HGNC:11132; resistance to inhibitors of cholinesterase 4 homolog; RIC4; RIC-4; SEC9; si:dkeyp-8f4.6; Snap; SNAP 25; snap25; SNAP-25; snap25.1; SNAP-25.1; snap25a; SNAP-25A; SNAP-25B; snp25; sp; SUP; super protein; synaptosomal associated protein-25; synaptosomal-associated 25 kDa protein; Synaptosomal-associated protein 25; synaptosomal-associated protein 25-A; synaptosomal-associated protein, 25 kDa; synaptosomal-associated protein, 25a; synaptosomal-associated protein, 25kDa; synaptosome associated protein 25; synaptosome associated protein 25a; synaptosome associated protein 25kDa; synaptosome-associated protein 25.1; synaptosome-associated protein 25a; wu:fj42b02; wu:fj45g04; wu:fj48c02 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
20614, 25012, 6616 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P60879, P60880, P60881 | |
Snap25 | |
Synthetic peptide corresponding to the C terminus amino acids DEANQRATKMLGSG. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |