Learn More
SHH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519492
Additional Details : Weight : 0.01000kg
Description
Not suitable for use with Immunohistochemistry - Free Floating. This antibody is predicted to react with rat, chicken, cow, human, Xenopus laevis, zebrafish and many species based on sequence homology.
SHH Polyclonal antibody specifically detects SHH in Human, Mouse samples. It is validated for ELISA, Western Blot.Specifications
SHH | |
Polyclonal | |
Unconjugated | |
Shh | |
9530036O11Rik; Dsh; fc83d08; hedgehog; hemimelic extra toes; Hhg1; HHG-1; HLP3; HPE3; Hx; Hxl3; M100081; MCOPCB5; morphogen; shh; Shh N-terminal processed signaling domains; Shh unprocessed N-terminal signaling and C-terminal autoprocessing domains; shh.L; shha; ShhN; ShhNC; ShhNp; short digits; SMMCI; sonic hedgehog; sonic hedgehog a; sonic hedgehog homolog; sonic hedgehog L homeolog; sonic hedgehog protein; Sonic hedgehog protein 19 kDa product; Sonic hedgehog protein A; Sonic hedgehog protein A N-product; Sonic hedgehog protein N-product; sonic hedgehog signaling molecule; sonic hedgehog signaling molecule a; sonic-you; syu; TPT; TPTPS; vhh1; vhh-1; wu:fc83d08; XELAEV_18030676mg; Xhh; xshh; X-SHH | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20423, 6469 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
0.7 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
Q15465, Q62226 | |
Shh | |
Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human Sonic Hedgehog. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |