missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535983
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: MCF-7. Predicted reactivity: Mouse (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SOX3 Polyclonal antibody specifically detects SOX3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
SOX3 | |
Polyclonal | |
Unconjugated | |
SOX3 | |
GHDX; LOC679158; MRGH; PHP; PHPX; similar to SRY (sex determining region Y)-box 3; Sox3; Sox-3; SOXB; SRY (sex determining region Y)-box 3; SRY box 3; SRY-box 3; SRY-box containing gene 3; transcription factor SOX-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20675, 6658, 679158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P41225, P53784 | |
SOX3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 159 of SOX3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |