Learn More
SOX4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541442
3815.09 SEK valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: STASTGGKAD DPSWCKTPSG HIKRPMNAFM VWSQIERRKI MEQSPDMHNA Sequence homology: Cow: 100%; Dog: 100%; Goat: 91%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
SOX4 Polyclonal antibody specifically detects SOX4 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
SOX4 | |
Polyclonal | |
Unconjugated | |
SOX4 | |
AA682046; ecotropic viral integration site 16; EVI16; SOX4; Sox-4; SRY (sex determining region Y)-box 4; SRY box 4; SRY-box 4; SRY-box containing gene 4; SRY-box transcription factor 4; SRY-related HMG-box gene 4; transcription factor SOX-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20677, 6659 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q06831, Q06945 | |
SOX4 | |
Synthetic peptide directed towards the N-terminal of human SOX4 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |