missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPHK1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518905
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
SPHK1 Polyclonal antibody specifically detects SPHK1 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
SPHK1 | |
Polyclonal | |
Unconjugated | |
SPHK1 | |
1110006G24Rik; Acetyltransferase SPHK1; RP23-193A16.4; SK 1; Sk1; SKase; Sphingosine kinase 1; SPHK; SPHK1; SPK; SPK 1; Spk1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8877 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9NYA1 | |
SPHK1 | |
Synthetic peptide sequence (DVDLESEKYRRLGE) corresponding to the internal amino acids of SPHK1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |