Learn More
SPR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522293
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2. Predicted reactivity: Rabbit (86%), Rhesus Monkey (97%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SPR Polyclonal antibody specifically detects SPR in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
SPR | |
Polyclonal | |
Unconjugated | |
SPR | |
AA409688; ENSMUSG00000071345; Gm10328; SDR38C1; sepiapterin reductase; sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase); short chain dehydrogenase/reductase family 38C, member 1; SPR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20751, 6697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P35270, Q64105 | |
SPR | |
Recombinant fragment corresponding to a region within amino acids 65 and 261 of SPR (Uniprot ID#P35270) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |