missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPRY4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526959
Additional Details : Weight : 0.01000kg
Description
SPRY4 is an inhibitor of the receptor-transduced mitogen-activated protein kinase (MAPK) signaling pathway. The protein is positioned upstream of RAS activation and impairs the formation of active GTP-RAS.Specifications
SPRY4 | |
Polyclonal | |
SPRY4 | |
protein sprouty homolog 4, spry-4 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
81848 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Q9C004 | |
SPRY4 | |
KLH conjugated synthetic peptide between 26-56 amino acids from human SPRY4 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |