missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRGAP2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519234
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology.
SRGAP2 Polyclonal antibody specifically detects SRGAP2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SRGAP2 | |
Polyclonal | |
Unconjugated | |
SRGAP2 | |
9930124L22Rik; AI448945; ARHGAP34; FBP2; Fbp27; FBP-27; FNBP2; formin binding protein 2; formin-binding protein 2; Formin-binding protein 27; KIAA0456; RGD1566016; rho GTPase-activating protein 34; SLIT-ROBO GAP2; SLIT-ROBO Rho GTPase activating protein 2; SLIT-ROBO Rho GTPase-activating protein 2; Srgap2; SRGAP2A; srGAP3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23380 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75044 | |
SRGAP2 | |
Synthetic peptide sequence (KQEDRQTPRSPDST) corresponding to the internal amino acids of SRGAP2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |