missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528259
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
STS Polyclonal antibody specifically detects STS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
STS | |
Polyclonal | |
Unconjugated | |
Sts | |
ArsC; ARSC1; Arylsulfatase C; ASC; ES; Estrone sulfatase; LOC402795 protein; LOW QUALITY PROTEIN: steryl-sulfatase; steryl-sulfatase; si:ch211-271l9.1; SSDD; Steroid sulfatase; steroid sulfatase (microsomal), arylsulfatase C, isozyme S; steroid sulfatase (microsomal), isozyme S; steroid sulphatase; steryl-sulfatase; Steryl-sulfate sulfohydrolase; STS; XLI | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
412 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P08842 | |
Sts | |
Recombinant fragment corresponding to a region within amino acids 96 and 547 of Human Steryl-sulfatase | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |