missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA112860
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is human brain lysate.
SULF2 Polyclonal antibody specifically detects SULF2 in Human samples. It is validated for Western Blot.Specifications
SULF2 | |
Polyclonal | |
Unconjugated | |
SULF2 | |
2010004N24Rik; AU020235; DKFZp313E091; Extracellular sulfatase Sulf-2; Extracellular sulfatase Sulf-2-like protein; FLJ44488; FLJ90554; HSULF-2; KIAA1247; MGC126411; mKIAA1247; mSulf-2; OTTHUMP00000031223; RP5-1049G16.1; SULF2; sulfatase 2; sulfatase FP2; UNQ559/PRO1120 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
55959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
Q8IWU5 | |
SULF2 | |
Synthetic peptide corresponding to human Sulf2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |