missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT4A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546732
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ELTSPRLIKS HLPYRFLPSD LHNGDSKVIY MARNPKDLVV SYYQFHRSLR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%
SULT4A1 Polyclonal antibody specifically detects SULT4A1 in Human samples. It is validated for Western Blot.Specifications
SULT4A1 | |
Polyclonal | |
Unconjugated | |
SULT4A1 | |
2400007A17Rik; AI853543; brain sulfotransferase 4A1; brain sulfotransferase-like protein; BRSTL1; BR-STL-1; DJ388M5.3; family 4A, member 1; hBR-STL; hBR-STL-1; LOW QUALITY PROTEIN: sulfotransferase 4A1; mBR-STL; nervous system cytosolic sulfotransferase; nervous system sulfotransferase; NST; rBR-STL; ST4A1; sulfotransferase 4A1; sulfotransferase family 4A member 1; sulfotransferase family 4A, member 1; sulfotransferase-related protein; SULT4A1; SULTX3; Unknown (protein for MGC:136828); zgc:136828 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25830 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BR01 | |
SULT4A1 | |
Synthetic peptide directed towards the middle region of human SULT4A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |