missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUMO2/SUMO3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519418
Additional Details : Weight : 0.01000kg
Description
Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with rat, cow, pig and zebrafish based on sequence homology.
SUMO2/SUMO3 Polyclonal antibody specifically detects SUMO2/SUMO3 in Hamster, Human, Mouse, X. laevis samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SUMO2/SUMO3 | |
Polyclonal | |
Unconjugated | |
Sumo3 | |
2810014B19Rik; D10Ertd345e; GMP1; HSMT3; I79_000932; MIF2 suppressor; Sentrin; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 3; small ubiquitin-like modifier 3 L homeolog; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 3; Smt3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 homolog 2; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); SMT3 supressor of mif two 3 homolog 2; SMT3A; Smt3B; SMT3C; SMT3H1; SMT3H2; SMT3H3; SUMO-1; SUMO2; SUMO-2; Sumo3; SUMO-3; sumo3.L; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; UBL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100689105, 100765708, 170930, 20610, 379449, 6612, 6613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.9 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
P55854, P61956, P61957, Q6LDZ8, Q7SZ22, Q9Z172 | |
SUMO2, Sumo3, sumo3.L | |
Synthetic peptide conjugated to KLH derived from within residues 50 to the C-terminus of Human Sumo 2+3. | |
100 μg | |
Primary | |
Hamster, Human, Mouse, X. laevis | |
Antibody | |
IgG |