Learn More
Superoxide Dismutase 4 Mouse anti-Human, Clone: 3A1, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Invitrogen LFMA0042
Additional Details : Weight : 0.99000kg
Product Code. 10292920
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
A suggested positive control for this product is HeLa cells.
Superoxide dismutase (SOD) is an antioxidant enzyme involved in the defense system against reactive oxygen species (ROS). SOD catalyzes the dismutation reaction of superoxide radical anion (O2) to hydrogen peroxide, which is then catalyzed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each of Cu and Zn ion per subunit. The manganese containing 80 kDa tetrameric enzyme SOD2, is located in the mitochondrial matrix in close proximity to a primary endogenous source of superoxide, the mitochondrial respiratory chain. SOD3 is a heparin-binding multimer of disulfide-linked dimers, primarily expressed in human lungs, vessel walls and airways. SOD4 is a copper chaperone for superoxide dismutase (CCS), which specifically delivers Cu to copper/zinc superoxide dismutase. CCS may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor.
Specifications
Specifications
Superoxide Dismutase 4
Monoclonal
1 mg/mL
CCS
CCS, copper chaperone for superoxide dismutase
Mouse
Protein G
RUO
9973
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
3A1
Unconjugated
O14618
CCS
Purified, recombit, human superoxide dismutase 4 protein expressed in E. coli.
100 μL
Primary
Human
Antibody
IgG2a