missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Monoclonal Antibody (H.987.7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515207
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other types of apoptotic inhibitor proteins.
Survivin Monoclonal antibody specifically detects Survivin in Human, Non-human Primate samples. It is validated for Western Blot.Specifications
Survivin | |
Monoclonal | |
Unconjugated | |
Birc5 | |
AAC-11; AP14; Api4; apoptosis inhibitor 4; Apoptosis inhibitor survivin; baculoviral IAP repeat containing 5; baculoviral IAP repeat-containing 5; baculoviral IAP repeat-containing 5 (survivin); baculoviral IAP repeat-containing protein 5; BIRC 5; BIRC5; EPR 1; EPR-1; Iap4; inhibitor of apoptosis; survivin; survivin variant 3 alpha; survivin40; SVV; TIAP | |
Mouse | |
Affinity chromatography | |
RUO | |
332 | |
-20°C | |
Liquid |
Western Blot | |
H.987.7 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
O15392 | |
Birc5 | |
Synthetic peptide corresponding to the amino-terminal sequence of human survivin | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |