Learn More
SUV39H1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529470
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, H1299, Raji, Mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (90%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SUV39H1 Polyclonal antibody specifically detects SUV39H1 in Human, Mouse, Non-human Primate samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
SUV39H1 | |
Polyclonal | |
Unconjugated | |
SUV39H1 | |
AI852103; AL022883; DXHXS7466e; H3-K9-HMTase 1; Histone H3-K9 methyltransferase 1; histone-lysine N-methyltransferase SUV39H1; histone-lysine N-methyltransferase, H3 lysine-9 specific 1; KMT1A; Lysine N-methyltransferase 1A; MG44; mIS6; Position-effect variegation 3-9 homolog; RGD1565028; Su(var)3-9 homolog 1; suppressor of variegation 3-9 homolog 1; suppressor of variegation 3-9 homolog 1 (Drosophila); SUV39H; Suv39h histone methyltransferase; SUV39H1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20937, 6839 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.52 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O43463, O54864 | |
SUV39H1 | |
Recombinant fragment corresponding to a region within amino acids 179 and 412 of Human SUV39H1 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG |