missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV39H2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518115
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine, human, mouse and porcine based on sequence homology.
SUV39H2 Polyclonal antibody specifically detects SUV39H2 in Rat samples. It is validated for Western Blot.Specifications
SUV39H2 | |
Polyclonal | |
Unconjugated | |
SUV39H2 | |
4930507K23Rik; AA536750; D030054H19Rik; D2Ertd544e; FLJ23414; H3 K9 HMTase 2; H3-K9-HMTase 2; Histone H3-K9 methyltransferase 2; histone-lysine N-methyltransferase SUV39H2; KMT1; KMT1B; lysine N-methyltransferase 1B; RP11-2K17.2; Su; Su(var)3-9 homolog 2; suppressor of variegation 3-9 homolog 2; suppressor of variegation 3-9 homolog 2 (Drosophila); Suv39h histone methyltransferase; Suv39h2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
364785 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
0 | |
SUV39H2 | |
Synthetic peptide sequence (CKCGAVTCRGYLN) corresponding to the C-terminus amino acids of SUV39H2 | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG |