Learn More
SWI6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14977
Additional Details : Weight : 0.01000kg
Description
PA1-4977 detects Swi6 from S. pombe samples. PA1-4977 has been successfully used in Western blot and immunoprecimmunoprecipitationitation applications. Western blot analysis of crude extract of S. pombe detects a protein with an apparent molecular size of 53 kD.
SWI6 Polyclonal antibody specifically detects SWI6 in Yeast samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.Specifications
SWI6 | |
Polyclonal | |
Unconjugated | |
SWI6 | |
Cell-cycle box factor subunit SWI6; Chromatin-associated protein swi6; chromodomain protein Swi6; L9470.8; MBF subunit P90; PSL8; Regulatory protein SWI6; SDS11; SPAC664.01c; SPAC824.10c; SWI6; Trans-acting activator of HO endonuclease gene; transcriptional regulator SWI6; YLR182W | |
Rabbit | |
50 μL | |
Primary | |
Yeast | |
Antibody | |
IgG |
ChIP Assay, Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum with 0.09% sodium azide | |
P09959 | |
SWI6 | |
Recombinant full length protein corresponding to Schizosaccharomyces pombe Swi6, aa 1-328. | |
RUO | |
850879 | |
-20°C | |
Liquid |