missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAMP1 Monoclonal Antibody (4E206), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA126000
Additional Details : Weight : 0.01000kg
Description
MA1-26000 detects a band of ∽38 kDa. Postive control of mouse and rat brain tissue extract suggested. No reactivity was detected in mouse liver extracts.
VAMP1 Monoclonal antibody specifically detects VAMP1 in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
VAMP1 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.2 | |
P23763, Q0V7N0, Q62442, Q63666 | |
Vamp1 | |
Synaptic vesicle-containing fractions, immunoprecipitated from human brain homogenates. | |
50 μL | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgM |
Immunohistochemistry, Western Blot | |
Clone: 4E206 | |
Unconjugated | |
Vamp1 | |
DKFZp686H12131; lew; MRXSYP; SPAX1; SYB1; Syb-1; synaptobrevin 1; synaptobrevin-1; vamp; Vamp1; VAMP-1; vesicle associated membrane protein 1; vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1) | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
22317, 25624, 513621, 6843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |