missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin 1 Monoclonal Antibody (ASV30), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125568
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Mouse or Rat brain tissue extract.. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Synaptotagmin 1 Monoclonal antibody specifically detects Synaptotagmin 1 in Bovine, Mouse, Rabbit, Rat, Shark, X. laevis samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
Synaptotagmin 1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1mM PMSF, 50% glycerol and no preservative; pH 7.2 | |
P21707, P46096, P48018 | |
Syt1, syt1.L | |
Rat brain synaptic junctional protein complexes. | |
100 μg | |
Primary | |
Bovine, Mouse, Rabbit, Rat, Shark, X. laevis | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
ASV30 | |
Unconjugated | |
Syt1 | |
AW124717; DKFZp781D2042; FLJ42519; G630098F17Rik; P65; SV DKFZp781D2042; SV SYT; svp65; Syb1; synaptotagmin 1; synaptotagmin 1 L homeolog; synaptotagmin 2; synaptotagmin I; synaptotagmin I protein; synaptotagmin I variant b; Synaptotagmin II; Synaptotagmin XI; synaptotagmin-1; Synaptotagmin-2; SYT; Syt1; syt1.L; SYT2; SytI; SytII; XELAEV_18022019mg | |
Mouse | |
Protein G | |
RUO | |
100350072, 20979, 25716, 281511, 447431 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |