missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSS00031W
Additional Details : Weight : 0.01000kg
Description
Glycerol (1:1) may be added for additional stability.
This gene encodes a protein that is a member of the syntaxin or t-SNARE (target-SNAP receptor) family. These proteins are found on cell membranes and serve as the targets for V-SNARES (vesicle-SNAP receptors) permitting specific synaptic vesicle docking and fusion. A microdeletion in the region of chromosome 20 where this gene is located has been associated with pseudohypoparathyroidism type Ib. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
STX16 | |
Polyclonal | |
STX16 | |
Syntaxin 16, STX16, Syn16 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
O14662, Q8BVI5 | |
STX16 | |
A synthetic peptide from aa region 100-200 of human Syntaxin-16 | |
RUO | |
228960, 8675 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |