Learn More
Syntaxin 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531208
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse brain, rat brain. Predicted reactivity: Mouse (99%), Rat (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Syntaxin 3 Polyclonal antibody specifically detects Syntaxin 3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Syntaxin 3 | |
Polyclonal | |
Unconjugated | |
Stx3 | |
STX3; STX3A; Syn-3; syntaxin 3; syntaxin 3A; syntaxin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20908, 6809, 81802 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.68 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q08849, Q13277, Q64704 | |
Stx3 | |
Recombinant fragment corresponding to a region within amino acids 1 and 214 of Human Syntaxin 3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |