missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540573
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SIPSPPGPNC QFLGGDHYSP LLPNQYPVPS RFYPDLPGQA KDVVPQAYWL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 86%; Rat: 86%
T-bet Polyclonal antibody specifically detects T-bet in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
T-bet | |
Polyclonal | |
Unconjugated | |
Tbx21 | |
HGNC:11599; LOW QUALITY PROTEIN: T-box transcription factor TBX21; Tbet; T-bet; TBLYM; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor 21; T-box transcription factor TBX21; TBT1; TBX21; T-cell-specific T-box transcription factor T-bet; T-PET; transcription factor TBLYM; transcription factor T-box 21 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
30009 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UL17 | |
Tbx21 | |
Synthetic peptide directed towards the middle region of human TBX21 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |