Learn More
Brachyury Monoclonal Antibody (1H9A2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517185
Additional Details : Weight : 0.01000kg
Description
MA5-17185 targets Brachyury protein T in indirect ELISA, FACS, IHC and WB applications and shows reactivity with Human samples. The MA5-17185 immunogen is purified recombinant fragment of human T (amino acids: 257-309 ) expressed in E. Coli. MA5-17185 detects Brachyury protein T which has a predicted molecular weight of approximately 47.4kDa.
Brachyury Monoclonal antibody specifically detects Brachyury in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Brachyury | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O15178 | |
TBXT | |
Purified recombinant fragment of human T (amino acids: 257-309 ) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
1H9A2 | |
Unconjugated | |
TBXT | |
Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3 | |
Mouse | |
Protein G | |
RUO | |
6862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |