missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACR1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518712
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
TACR1 Polyclonal antibody specifically detects TACR1 in Human, Mouse samples. It is validated for Western Blot.Specifications
TACR1 | |
Polyclonal | |
Unconjugated | |
Tacr1 | |
neurokinin 1 receptor; neurokinin receptor 1; neurokinin receptor subtype NK1; neurokinin-1 receptor; Nk1 receptor; NK-1 receptor; Nk1r; NK-1R; NKIR; SPR; substance p receptor; substance-P receptor; TAC1R; tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor); TACR1; truncated neurokinin-1 receptor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
21336, 6869 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P25103, P30548 | |
Tacr1 | |
Synthetic peptide sequence (EIPGDSSDRYHEQ) corresponding to the internal amino acids of TACR1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |