Learn More
TADA3L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540382
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ASVEPYCADI TSEEVRTLEE LLKPPEDEAE HYKIPPLGKH YSQRWAQEDL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%
TADA3L Polyclonal antibody specifically detects TADA3L in Human samples. It is validated for Western Blot.Specifications
TADA3L | |
Polyclonal | |
Unconjugated | |
TADA3 | |
1110004B19Rik; ADA3; ADA3 homolog; ADA3-like protein; AI987856; alteration/deficiency in activation 3; fa03f04; FLJ20221; FLJ21329; hADA3; mADA3; NGG1; STAF54; Tada3; Tada3l; Transcriptional adapter 3; transcriptional adapter 3-like; transcriptional adaptor 3; transcriptional adaptor 3 (NGG1 homolog, yeast)-like; transcriptional adaptor 3-like isoform b; Unknown (protein for MGC:133915); wu:fa03f04; zgc:63596 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10474 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O75528 | |
TADA3 | |
Synthetic peptide directed towards the middle region of human TADA3L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |