missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAFA5 Monoclonal Antibody (463102), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA524183
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA2, 3, or 4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
TAFA5 Monoclonal antibody specifically detects TAFA5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TAFA5 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q7Z5A7 | |
TAFA5 | |
E. coli-derived recombinant human TAFA5/FAM19A5 Gln26-Ser125 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
463102 | |
Unconjugated | |
TAFA5 | |
Chemokine-like protein TAFA-5; FAM19A5; family with sequence similarity 19 (chemokine (C-C motif)-like), member A5; family with sequence similarity 19 member A5, C-C motif chemokine like; family with sequence similarity 19, member A5; protein FAM19A5; QLLK5208; RGD1562115; TAFA protein 5; TAFA5; Tafa-5; Tara-5; UNQ5208; UNQ5208/PRO34524 | |
Rat | |
Protein A/G | |
RUO | |
25817 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |