missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carboxypeptidase B2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA143056
Additional Details : Weight : 0.01000kg
Description
PA1-43056 detects TAFI in human samples. PA1-43056 has successfully been used in Western blot and ELISA applications. PA1-43056 was derived from purified human TAFI.
Carboxypeptidase B2 Polyclonal antibody specifically detects Carboxypeptidase B2 in Human samples. It is validated for ELISA, Western Blot.Specifications
Carboxypeptidase B2 | |
Polyclonal | |
Unconjugated | |
CPB2 | |
1110032P04Rik; 4930405E17Rik; AI255929; carboxypeptidase B2; carboxypeptidase B2 (plasma); carboxypeptidase B2 (plasma, carboxypeptidase U); carboxypeptidase B-like protein; carboxypeptidase R; Carboxypeptidase U; Cpb2; CPR; CPU; pCPB; Plasma carboxypeptidase B; Tafi; thrombin-activable fibrinolysis inhibitor; thrombin-activatable fibrinolysis inhibitor | |
Sheep | |
Ammonium sulfate precipitation, Ion-exchange chromatography | |
RUO | |
1361 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
26 mg/mL | |
50% water with 50% glycerol and no preservative | |
Q96IY4 | |
CPB2 | |
Purified Human TAFI. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG |