missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAOK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534250
Additional Details : Weight : 0.01000kg
Description
TAOK1 Polyclonal antibody specifically detects TAOK1 in Bovine, Canine, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rat, Xenopus laevis samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TAOK1 | |
Polyclonal | |
Unconjugated | |
TAOK1 | |
2810468K05Rik; AU020252; D130018F14Rik; FLJ14314; h hTAOK1; H KIAA1361; hKFC-B; hTAOK1; KFC-B; Kiaa1361; Kinase from chicken homolog B; MAP3K16; MARK Kinase; Markk; microtubule affinity regulating kinase kinase; mKIAA1361; prostate-derived STE20-like kinase 2; prostate-derived sterile 20-like kinase 2; PSK2; PSK-2; serine/threonine protein kinase TAO1; serine/threonine protein kinase TAO1 homolog; serine/threonine-protein kinase TAO1; STE20-like kinase PSK2; tao; TAO kinase 1; TAO1; TAOK1; Thousand and one amino acid protein 1; thousand and one amino acid protein kinase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100059705, 100622172, 100758772, 216965, 286993, 480632, 536532, 57551 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O88664, Q5F2E8, Q7L7X3 | |
TAOK1 | |
Synthetic 16 amino acid peptide from internal region of human TAOK1. | |
50 μg | |
Primary | |
Bovine, Canine, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rat, X. laevis | |
Antibody | |
IgG |