missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ TAOK2 Polyclonal Antibody

Beskrivning
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (93%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TAOK2 (TAO1) is a serine/threonine kinase that may be an important regulator of the stress-responsive p38 MAP kinase pathway. In vitro, TAO (thousand and one amino acid) protein kinase 2 (TAO2) activates MAP/ERK kinases (MEK ) 3, 4, and 6 toward their substrates p38 MAP kinase JNK/SAPK (Chen et al., 1999; Chen and Cobb, 2001). This and more recent work has led to the proposal that the TAO protein kinases play an essential role in signaling from physiological agonist to the stress -responsive p38 MAPK (Chen et al., 2003). Autophosphorylation of TAO may play a role in the mechanism of TAO activation. The MEK binding domain of TAO is autophosphorylated on both serine and threonine residues and Ser181 is located within this domain.
Specifikationer
Specifikationer
| Antigen | TAOK2 |
| Användningsområden | Western Blot, Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gen | TAOK2 |
| Genaccessionsnr. | Q6ZQ29, Q9UL54 |
| Gene Alias | 1110033K02Rik; B230344N16; hKFC-C; Kiaa0881; Kinase from chicken homolog C; MAP3K17; mKIAA0881; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; prostate-derived sterile 20-like kinase 1; PSK; PSK1; PSK-1; PSK1-BETA; serine/threonine protein kinase TAO2; serine/threonine-protein kinase TAO2; serine-threonine kinase; tao; TAO kinase 2; TAO kinase 2TAO kinase 2; TAO1; TAO2; TAOK2; Thousand and one amino acid protein 2; thousand and one amino acid protein kinase 2; UNQ2971/PRO7431 |
| Gensymboler | TAOK2 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?