missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518850
Additional Details : Weight : 0.01000kg
Description
TAS1R2 Polyclonal antibody specifically detects TAS1R2 in Human samples. It is validated for Western Blot.Specifications
TAS1R2 | |
Polyclonal | |
Unconjugated | |
TAS1R2 | |
aldehyde dehydrogenase 4 family, member A1; Aldh4a1; candidate taste receptor T1R2; G protein-coupled receptor 71; GPCRB4; GPR71; G-protein coupled receptor 71; Sweet taste receptor T1R2; T1R2; Tas1r2; taste 1 receptor member 2; taste receptor TR2; taste receptor type 1 member 2; taste receptor, type 1, member 2; Tr2; TS1R2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
80834 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8TE23 | |
TAS1R2 | |
Synthetic peptide sequence (CSKRCQSGQKKKP) corresponding to the internal amino acids of TAS1R2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |