missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AS160 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534524
Additional Details : Weight : 0.01000kg
Description
PA5-34524 can be used with blocking peptide PEP-1567.
AS160 Polyclonal antibody specifically detects AS160 in Human samples. It is validated for Immunohistochemistry.Specifications
AS160 | |
Polyclonal | |
Unconjugated | |
TBC1D4 | |
5930406J04Rik; A930035N22; Akt substrate of 160 kDa; AS160; AV295684; DKFZp779C0666; Kiaa0603; NIDDM5; OTTHUMP00000040818; RGD1561609; RP11-159J2.1; RP11-159J2.3; similar to TBC1 domain family member 4; TBC (Tre-2, BUB2, CDC16) domain-containing protein; TBC Tre-2; TBC1 domain family member 4; TBC1 domain family, member 4; TBC1D4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9882 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O60343 | |
TBC1D4 | |
An 19 amino acid peptide near the carboxy terminus of human TBC1D4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |