missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517478
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
TBK1 Polyclonal antibody specifically detects TBK1 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
TBK1 | |
Polyclonal | |
Unconjugated | |
TBK1 | |
1200008B05Rik; AI462036; AW048562; FLJ11330; FTDALS4; NAK; NF-kappa-B-activating kinase; NF-kB-activating kinase; serine/threonine protein kinase TBK1; serine/threonine-protein kinase TBK1; T2K; TANK binding kinase 1; TANK-binding kinase 1; tbk; TBK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29110, 299827, 56480 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
174 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q9UHD2, Q9WUN2 | |
TBK1 | |
Synthetic peptide corresponding to residues near the carboxy-terminus of TBK1/NAK | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |