missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX22 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538836
Additional Details : Weight : 0.00900kg
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. Mutations in this gene have been associated with the inherited X-linked disorder, Cleft palate with ankyloglossia, and it is believed to play a major role in human palatogenesis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Jul 2008].Specifications
TBX22 | |
Polyclonal | |
Unconjugated | |
TBX22 | |
TBOX22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50945 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9Y458 | |
TBX22 | |
A synthetic peptide derived from the N-terminal region of human TBX22 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |