missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539499
Additional Details : Weight : 0.00900kg
Description
This gene encodes a member of the T cell factor/lymphoid enhancer factor family of transcription factors. These transcription factors are activated by beta catenin, mediate the Wnt signaling pathway and are antagonized by the transforming growth factor beta signaling pathway. The encoded protein contains a high mobility group-box DNA binding domain and participates in the regulation of cell cycle genes and cellular senescence. [provided by RefSeq, Nov 2010].Specifications
TCF7L1 | |
Polyclonal | |
Unconjugated | |
TCF7L1 | |
TCF3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21415, 83439 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9HCS4, Q9Z1J1 | |
TCF7L1 | |
A synthetic peptide derived from the internal region of human TCF7L1 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |