missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP-1 zeta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537073
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 60 kDa. Purity is >95% by SDS-PAGE.
TCP-1 zeta Polyclonal antibody specifically detects TCP-1 zeta in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TCP-1 zeta | |
Polyclonal | |
Unconjugated | |
CCT6A | |
acute morphine dependence related protein 2; acute morphine dependence-related protein 2; amino acid transport defect-complementing; CCT zeta1; CCT- zeta1; CCT- zeta-1; CCT zeta-1; Cct6; Cct6a; Cctz; Cctz1; Cctz-1; CCT-zeta; CCT-zeta-1; chaperonin containing T-complex subunit 6; chaperonin containing TCP-1; chaperonin containing TCP1 subunit 6A; chaperonin containing Tcp1, subunit 6A (zeta 1); chaperonin containing Tcp1, subunit 6a (zeta); chaperonin subunit 6a (zeta); histidine transp; histidine transport regulator 3; HTR3; MoDP-2; T-complex protein 1 subunit zeta; T-complex protein 1, zeta subunit; TCP 1 zeta; TCP1 zeta; TCP-1- zeta; TCP1- zeta; TCP-1-zeta; Tcp20; TCPZ; TTCP20 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12466, 288620, 908 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P40227, P80317 | |
CCT6A | |
Synthetic peptide corresponding to amino acids 216-261 of human TCP-1 zeta | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |