Learn More
TCR beta Monoclonal Antibody (H57-597), Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen MA170045
Additional Details : Weight : 0.01000kg
Description
MA1-70045 detects alpha/beta TCR in mouse samples. MA1-70045 has been successfully used in flow cytometry, immunoprecipitation, immunohistochemistry (frozen sections), and Western blot procedures. The MA1-70045 immunogen is affinity purified D0 -11.10 TCR. Stable at 4oC. For long term storage, aliquot and freeze unused portions at -20oC in volumes appropriate for single usage. Avoid repeated freeze/thaw cycles.
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
TCR beta | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
0 | |
Tcrb | |
Affinity purified D0 -11.10 TCR. | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
H57-597 | |
Unconjugated | |
Tcrb | |
bTCR; FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; T cell receptor beta chain; t-cell antigen receptor; T-cell receptor; TCR beta; Tcra; Tcrb; TCRbeta; TCRD; Tib; TRA | |
Armenian Hamster | |
Protein G | |
RUO | |
21577 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |