missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTEX1D3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534446
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is EL4 cell lysate. PA5-34446 can be used with blocking peptide PEP-1488.
TCTEX1D3 Polyclonal antibody specifically detects TCTEX1D3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
TCTEX1D3 | |
Polyclonal | |
Unconjugated | |
TCTE3 | |
D17Leh117c; LC2; t-complex testis-expressed 4; T-complex testis-specific protein 2; T-complex testis-specific protein 3; t-complex-associated testis expressed 3; T-complex-associated testis-expressed protein 3; T-complex-associated testis-expressed protein 4; t-complex-associated-testis-expressed 3; TCTE3; tcte-3; Tctex1 domain containing 3; tctex1 domain-containing protein 3; Tctex1d3; Tctex2; TCTEX-2; Tctex4; Tctex-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21647, 365153, 6991 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P11985, Q8IZS6 | |
TCTE3 | |
A 19 amino acid synthetic peptide near the amino terminus of human TCTEX1D3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |