missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cripto Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534476
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is 293 cell lysate. PA5-34476 can be used with blocking peptide PEP-1518.
Cripto Polyclonal antibody specifically detects Cripto in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Cripto | |
Polyclonal | |
Unconjugated | |
TDGF1 | |
CR; CR1; CRGF; CRIPTO; cripto growth factor; Cripto-1; cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; epidermal growth factor-like cripto protein CR1; Tdgf1; TDGF2; teratocarcinoma-derived growth factor; teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21667, 680246, 6997 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P13385, P51865 | |
TDGF1 | |
A 17 amino acid peptide near the amino terminus of human TDGF1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |