Läs mer
Invitrogen™ TGN46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1-1069
Beskrivning
PA1-1069 detects TGN46 in human samples. PA1-1069 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects a ~51 kDa protein representing TGN46 in HEK293 and HeLa cell lysates. The PA1-1069 immunogen is a synthetic peptide corresponding to residues V(252) V P E Q P S W K D H S K P I S(267) of human TGN46. This peptide (Cat. # PEP-291) is available for use in neutralization and control experiments.
IASPP is one of the most evolutionarily conserved inhibitors of p53 (TP53; MIM 191170), whereas ASPP1. and ASPP2. are activators of p53.
Specifikationer
| TGN46 | |
| Polyclonal | |
| Unconjugated | |
| Tgoln2 | |
| AU015504; D6Ertd384e; hTGN48; hTGN51; Tgn38; TGN38 homolog; TGN38A; TGN38B; TGN41; TGN46; TGN48; TGN51; Tgoln1; Tgoln2; Trans-Golgi network glycoprotein 46; Trans-Golgi network glycoprotein 48; Trans-Golgi network glycoprotein 51; Trans-Golgi network integral membrane protein 1; trans-Golgi network integral membrane protein 2; Trans-Golgi network integral membrane protein TGN38; trans-golgi network protein; trans-Golgi network protein (46, 48, 51kD isoforms); trans-golgi network protein 1; trans-golgi network protein 2; trans-Golgi network protein TGN51; Ttgn1; TTGN2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10618 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| O43493 | |
| Tgoln2 | |
| Synthetic peptide corresponding to residues V(252) V P E Q P S W K D H S K P I S(267) of human TGN46. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |