missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ TSPAN3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525472
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is predicted to react with bovine based on sequence homology.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. Two alternative transcripts encoding different isoforms have been described.
Specifikationer
| TSPAN3 | |
| Polyclonal | |
| Unconjugated | |
| TSPAN3 | |
| 1700055K04Rik; H41D4 protein; OAP-1; OSP-associated protein 1; tetraspan 3; tetraspan TM4SF; tetraspanin 3; tetraspanin TM4-A; tetraspanin TM4-A homolog; tetraspanin-3; TM4-A; Tm4sf8; Transmembrane 4 superfamily member 8; TSPAN3; TSPAN-3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10099, 56434 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O60637, Q9QY33 | |
| TSPAN3 | |
| KLH conjugated synthetic peptide between 90-117 amino acids from the central region of human TSPAN3. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering