missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546736
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: HTLGLSALNL DSGTEMPALT SSQMPSLSVL SVCTEESSPP NTGPTVTPPN
UBAP1 Polyclonal antibody specifically detects UBAP1 in Human samples. It is validated for Western Blot.Specifications
UBAP1 | |
Polyclonal | |
Unconjugated | |
UBAP1 | |
2700092A01Rik; NAG20; nasopharyngeal carcinoma-associated gene 20 protein; UAP; Ubap; Ubap1; UBAP-1; ubiquitin associated protein 1; ubiquitin-associated protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51271 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZ09 | |
UBAP1 | |
Synthetic peptide directed towards the C-terminal of human UBAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |